Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Rat anti-Mouse, Alexa Fluor 700, Clone: 910019, R&D Systems™

Rat MonoclonalAntibody

Manufacturer:  R&D Systems FAB8547N100UG

Catalog No. FAB8547N100


Add to cart
This item is not returnable. View return policy

Description

Description

Recombinant Monoclonal Antibody.
Specifications

Specifications

CD24
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P24807
CD24
Peptide sequence spanning a portion of the extracellular region Accession # P24807
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects mouse CD24a in direct ELISAs.
Flow Cytometry
910019
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD 24, CD24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24A, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Rat
IgG2a
RUO
Primary
100133941
Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)