Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD244.1 Rat anti-Mouse, APC, Clone: C9.1, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 17244082

 View more versions of this product

Catalog No. 50-112-3112


Add to cart

Description

Description

Description: The monoclonal antibody C9.1 reacts with mouse CD244.1. This receptor is expressed on NK cells, gamma delta T cells, some CD8+ T cells, basophils, and monocytes on most inbred mouse strains except C57BL/6 and C58, which express the CD244.2 allotype. CD244 is a CD2-related receptor belonging to the SLAM family that exists as two different isoforms with opposing functions. CD244 associates with CD48, which is expressed widely on hematopoietic cells, regulates NK cell proliferation, cytotoxicity, and cytokine production. However, the CD244/CD28 interaction between NK and other cell types inhibits NK cytotoxicity, which contributes to self-tolerance. CD244 may enhance or inhibit CD8+ T cell effector function depending on SAP levels. The C9.1 antibody has been reported to have antagonist activity on activated NK cells. Applications Reported: This C9.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This C9.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

Filtration: 0.2 µm post-manufacturing filtered. This gene encodes a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via this receptor is thought to modulate NK-cell cytolytic activity. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CD244.1
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q07763
Cd244
IgG2b, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
18106
Flow Cytometry
C9.1
APC
Cd244
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.