Learn More
CD244 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166771AP
Catalog No. 50-173-1106
Add to cart
Description
Description
CD244 (2B4, SLAMF4) is a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via CD244 is thought to modulate NK-cell cytolytic activity. CD244 interacts with SLAMF2, causing the activation of both SLAMF4- and SLAMF2-expressing cells. Patients with systemic lupus erythmatosus have lower than normal levels of SLAMF4 expressed on their NK cells and monocytes, suggesting that SLAMF4 may play a role in the pathology of this autoimmune disease. Diseases associated with CD244 dysfunction include rheumatoid arthritis and lymphoproliferative syndrome. Alternatively spliced transcript variants encoding different isoforms of CD244 have been found for this gene.
Specifications
Specifications
CD244
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BZW8
2B4, CD244, h2B4, NAIL, NKR2B4, SLAMF4
Rabbit
IgG
150 μL
-20°C
Primary
51744
Immunohistochemistry (Paraffin)
Unconjugated
CD244
Liquid
CD244
CD244 Fusion Protein Ag9860
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human