Learn More
BD Cell Analysis 3P CD25
Supplier:  BD Cell Analysis 3P 335807
Encompass
CD25 PE-Cy™7 Clone 2A3 (ASR), 100 Tests CD25, clone 2A3, is derived from hybridization of mouse myeloma cell line NS-1 myeloma cells with spleen cells from BALB/c mice immunized with PHA-activated human T lymphocytes. CD25 (Anti-IL-2R) recognizes an antigen that is the human low-affinity interleukin-2 receptor (IL-2R). The antigen has a molecular weight of 55 kilodaltons (kd). Concentration: 12.5 μg/mLVol. Per Test: 5 μL Isotype: Mouse BALB/c IgG1, κReactivity Human (QC Testing) Application: Flow cytometry (Routinely Tested) Immunogen: Human Phytohemagglutinin-activated T Cells Workshop No.: III A769,T153; IV A8 Storage Buffer: Phosphate buffered saline with gelatin and 0.1% sodium azide. Regulatory Status: ASR. 26628-22-8
Catalog No. 50-620-028
Due to product restrictions, please to purchase or view availability for this product.