Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Human anti-Human, Alexa Fluor 350, Clone: Hu102, R&D Systems
Human Monoclonal Antibody
Supplier: R&D Systems FAB9927U100UG
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide. | |
Human | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human CD25/IL-2 R alpha in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Daclizumab. This product is for research use only. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
Hu102 | |
Flow Cytometry 0.25-1 μg/10^6 cells | |
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
Human CD25/IL-2 R alpha | |
100 μg | |
3559 | |
Human | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title