Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2R alpha Human anti-Human, Cynomolgus Monkey, Rhesus Macaque, DyLight 755, Clone: Daclizumab, Novus Biologicals™

Human Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275901IR

Catalog No. NBP275901IR


Add to cart

Description

Description

CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD25/IL-2R alpha
Daclizumab
Flow Cytometry
Conjugated, Purified
IL2RA
Cultured T-cells derived from a patient with Mycosis fungoides.
Protein A purified
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary
3559.0
Cynomolgus Monkey, Human, Rhesus Macaque
Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Human
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit