Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2 R alpha Rat anti-Mouse, Clone: 280401, Novus Biologicals™
Rat Monoclonal Antibody
$175.82 - $436.00
Specifications
Antigen | CD25/IL-2R alpha |
---|---|
Clone | 280401 |
Concentration | 0.51 mg/mL |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB24381
|
Novus Biologicals
MAB24381 |
100 μg |
Each for $436.00
|
|
MAB24381SP
|
Novus Biologicals
MAB24381SP |
25 μg |
Each for $175.82
|
|
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
0.51 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
PBS with No Preservative | |
3559 | |
Partial recombinant mouse CD25/IL2Ra protein (amino acids 22-236) [UniProt P01590]. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
280401 | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
IL2RA | |
IgG2a | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title