Learn More
CD25/IL-2 R alpha Rat anti-Mouse, Clone: AP-MAB0710, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11097867100UG
Catalog No. NBT97867100
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD25/IL-2 R alpha
AP-MAB0710
Flow Cytometry 1:100-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20-1:300, CyTOF-ready
P01590
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Rat
IgG1
100 ug
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.4 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
A 0.2 um filtered solution in PBS with No Preservative
Purified
IL2RA
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with recombinant mouse CD25 protein.
Protein G purified
Adaptive Immunity, Apoptosis, Cytokine Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary
3559
Mouse