Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Rat anti-Mouse, Janelia Fluor 549, Clone: PC61, Novus Biologicals
Rat Monoclonal Antibody
Manufacturer: Novus Biologicals NBP227425JF549
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.Specifications
CD25/IL-2R alpha | |
PC61 | |
Flow Cytometry | |
Conjugated, Purified | |
IL2RA | |
The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody. | |
Protein G purified | |
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
Primary | |
3559.0 |
Flow Cytometry | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
Rat | |
IgG1 Lambda | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title