Learn More
CD25/IL-2R alpha Rabbit anti-Human, Mouse, Several Species, Alexa Fluor™ 594, Clone: Basiliximab, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252661AF594
Catalog No. NP22661A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Human,Mouse,Rat,Cynomolgus Monkey,Rhesus Macaque samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
CD25/IL-2R alpha
Basiliximab
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Basiliximab recognises the human IL-2R and binds to the epitope (116)ERIYHFV(122) within the extracellular domain of the CD25 subunit -binding is abolished when 2 or more residues within the epitope are mutated. This epitope overlaps with the interaction site of CD25 and IL-2. CD25 is found in the high-affinity IL-2R ((alpha)(beta)(gamma)c) -T cells only express CD25 upon activation. IL-2 binding to IL-2R on T cells modulates proliferation and differentiation.
Flow Cytometry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
IL2RA
Basiliximab was prepared by immunizing BALB/c mice with CTC 16 cells.
0.1 mL
Primary
3559
Cynomolgus Monkey, Human, Mouse, Rat, Rhesus Macaque