Learn More
CD25/IL-2R alpha Rat anti-Mouse, Alexa Fluor™ 350, Clone: PC61, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP227425AF350
Catalog No. NP22742A30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD25/IL-2R alpha
PC61
Flow Cytometry, Flow (Cell Surface)
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Rat
IgG1 Lambda
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
IL2RA
The B6.1 mouse cytotoxic T cell line was used as immunogen for this antibody.
0.1 mL
Primary
3559