Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: M-A251, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566448
Description
CD25 Monoclonal antibody specifically detects CD25 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
CD25 | |
Monoclonal | |
BB700 | |
25 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
M-A251 | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title