Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25 Mouse anti-Rat, FITC, Clone: OX-39, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 561783

 View more versions of this product

Catalog No. BDB561783


Add to cart

Description

Description

The OX-39 antibody reacts with the α chain of the IL-2 receptor on T lymphoblasts and thymic and splenic dendritic cells. CD25 has also been detected on rat intestinal epithelial cells. It has been reported that OX-39 mAb weakly blocks binding of IL-2 to T-cell blasts and that it blocks IL-2 stimulated epithelial cell migration in an in vitro model of wound healing.

Specifications

Specifications

CD25
OX-39
FITC
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Rat
Flow Cytometry
0.5mg/mL
IL-2R α Chain
Affinity Purified
Rat T blasts from mixed-lymphocyte reactions
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.