Learn More
CD252 Rat anti-Mouse, BB515, Clone: RM134L, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566213
Catalog No. BDB566213
Add to cart
Description
Description
CD252 Monoclonal antibody specifically detects CD252 in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
CD252
RM134L
BB515
Purified
Rat
Rat SD, also known as Sprague-Dawley (outbred) IgG2b, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Tnfsf4; OX40 ligand; OX40L; TXGP1; Txgp1l; gp34; Ath-1, Ath1, CD134L
Rat NRK-52E cell line transfected with mouse OX-40L gene (Txgp1l)
25μg
Cell Biology
Primary
Murine
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.