Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD254 (RANK Ligand) Mouse anti-Human, Clone: MIH24, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14661982

 View more versions of this product

Catalog No. 50-112-2711


Add to cart

Description

Description

Description: The MIH24 monoclonal antibody reacts with human TRANCE (TNF-related activation-induced cytokine receptor) also known as RANKL (receptor activator of NF-kappaB ligand) and OPGL (Osteoprotegerin Ligand). TRANCE is a type II membrane protein with predicted molecular weight of 35 kDa. Its extracellular domain is most closely related to TRAIL, FasL and TNF. TRANCE mRNA is upregulated by TCR stimulation. TRANCE-deficient mouse lacks osteoclasts. In addition, TRANCE has been shown to be involved in the interactions between T-dendritic and T-B cells. It was also found to be critical in osteoclast differentiation. The TRANCE-RANK interaction regulates the expression of the anti-apoptotic molecule Bcl-xL. Applications Reported: The MIH24 antibody has been reported for use in flow cytometric analysis, and ELISA. Applications Tested: The MIH24 antibody has been tested by flow cytometric analysis of human RANKL transfected cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.

2 µm post- manufacturing filtered. This gene encodes a member of the tumor necrosis factor (TNF) cytokine family which is a ligand for osteoprotegerin and functions as a key factor for osteoclast differentiation and activation. This protein was shown to be a dentritic cell survival factor and is involved in the regulation of T cell-dependent immune response. T cell activation was reported to induce expression of this gene and lead to an increase of osteoclastogenesis and bone loss. This protein was shown to activate antiapoptotic kinase AKT/PKB through a signaling complex involving SRC kinase and tumor necrosis factor receptor-associated factor (TRAF) 6, which indicated this protein may have a role in the regulation of cell apoptosis. Targeted disruption of the related gene in mice led to severe osteopetrosis and a lack of osteoclasts. The deficient mice exhibited defects in early differentiation of T and B lymphocytes, and failed to form lobulo-alveolar mammary structures during pregcy. Two alternatively spliced transcript variants have been found.
Specifications

Specifications

CD254 (RANK Ligand)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O14788
RP11-86N24.2, CD254, ODF, OPGL, OPTB2, RANKL, TRANCE, hRANKL2, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCE
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISA, Flow Cytometry
MIH24
Unconjugated
TNFSF11
Liquid
TNFSF11
IgG2b, kappa
100 μg
4° C
Primary
8600
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.