Learn More
CD26 Mouse anti-Human, APC, Clone: BA5b, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen CD2605
Catalog No. 50-112-1604
Add to cart
Description
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti CD3-induced IL - 2 production. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of maligt transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
Specifications
Specifications
CD26
Monoclonal
APC
DPP4
Liquid
DPP4
Human CD26
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
BA5b
PBS with BSA, sucrose and 0.1% sodium azide
P27487
ADABP, ADCP2, CD26, DPP-IV, TP103, ADCP-2, DPP IV, T-cell activation antigen CD26, adenosine deaminase complexing protein 2, dipeptidyl peptidase 4, dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidylpeptidase IV CD26, adenosine deaminase complexing protein 2
Mouse
IgG2a, kappa
0.5 mL
4° C, store in dark
Primary
1803