Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD26 Mouse anti-Human, PE, Clone: 2A6, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 12026941

 View more versions of this product

Catalog No. 50-112-2221


Add to cart

Description

Description

Description: The 2A6 monoclonal antibody reacts with human CD26, a 110 kDa type II transmembrane dimer. CD26 is an ectoenzyme with dipeptidyl peptidase IV activity expressed by T cells and is upregulated upon T cell stimulation. It is thought that CD26 plays a role in interaction with the extracellular matrix molecules and in T cell activation. Applications Reported: The 2A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2A6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD26 (dipeptidyl peptidase IV, DPP IV), adenosine deaminase (ADA) binding protein) is a homodimeric atypical serine protease belonging to the prolyl oligopeptidase family. CD26 is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti CD3-induced IL - 2 production. CD26 identical to adenosine deaminase complexing protein-2, and to the T-cell activation antigen CD26. Further, CD26 is an intrinsic membrane glycoprotein and a serine exopeptidase that cleaves X-proline dipeptides from the N-terminus of polypeptides. Alterations in CD26 peptidase activity are characteristic of maligt transformation, and the enzymatic activity increases dramatically with tumor grade and severity. CD26 is expressed in various blood cell types, but also in cells that are histogenetically related to activated fibroblasts. Alterations in CD26 density have been reported on circulating monocytes and CD4+ T cells during rheumatoid arthritis and systemic lupus erythematosus.
Specifications

Specifications

CD26
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P27487
ADABP, ADCP2, CD26, DPP-IV, TP103, ADCP-2, DPP IV, T-cell activation antigen CD26, adenosine deaminase complexing protein 2, dipeptidyl peptidase 4, dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidylpeptidase IV CD26, adenosine deaminase complexing protein 2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
2A6
PE
DPP4
Liquid
DPP4
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1803
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.