Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD26 Mouse anti-Rat, BUV615, Clone: OX-61, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752344
Description
CD26 Monoclonal antibody specifically detects CD26 in Rat samples. It is validated for Flow Cytometry.
Specifications
CD26 | |
Monoclonal | |
0.2 mg/mL | |
Rat CD26 BUV615 OX-61 50 μg | |
Rat dendritic cells enriched from thoracic duct lymph | |
RUO | |
CD26 | |
Refrigerate (2 to 8°C) | |
Rat |
Flow Cytometry | |
OX-61 | |
Brilliant Ultraviolet 615 | |
Mouse | |
1 x 50 μg | |
Primary | |
Rat | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction