Learn More
CD262 (TRAIL-R2) Mouse anti-Human, Alexa Fluor 647, Clone: B-K29, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565498
Catalog No. BDB565498
Add to cart
Description
Description
CD262 (TRAIL-R2) Monoclonal antibody specifically detects CD262 (TRAIL-R2) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD262 (TRAIL-R2)
B-K29
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
CD262; TNFRSF10B; TRAILR2; TRAIL-R2; DR5; TRICK2; ZTNFR9; KILLER
Human
50 Tests
Cell Biology
Primary
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.