Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD265/RANK Mouse anti-Human, Unconjugated, Clone: 8G1-24, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2074100UG
Description
CD265/RANK Monoclonal antibody specifically detects CD265/RANK in Human samples. It is validated for Western Blotting, Flow Cytometry.Specifications
CD265/RANK | |
Monoclonal | |
Unconjugated | |
Tumor necrosis factor receptor superfamily member 11A;Osteoclast differentiation factor receptor;ODFR;Receptor activator of NF-KB;CD265 | |
Mouse | |
66.03 kDa calculated. | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry | |
8G1-24 | |
NP_001257878 | |
TNFRSF11A, RANK | |
Extracellular domain of human RANK (CD265). | |
Protein G Purified | |
Inflammation and Immunology | |
Clone 8G1-24 is a mouse monoclonal antibody that detects human RANK(CD2 65). It targets an epitope within the extracellular domain. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction