Learn More
CD265/RANK Mouse anti-Human, Unconjugated, Clone: 8G1-24, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma MABS207425UG
 View more versions of this product
Catalog No. MABS20742MI
Add to cart
Description
Description
CD265/RANK Monoclonal antibody specifically detects CD265/RANK in Human samples. It is validated for Western Blotting, Flow Cytometry.
Specifications
Specifications
CD265/RANK
8G1-24
NP_001257878
Tumor necrosis factor receptor superfamily member 11A;Osteoclast differentiation factor receptor;ODFR;Receptor activator of NF-KB;CD265
Mouse
IgG1κ
Protein G Purified
Inflammation and Immunology
Primary
Human
Western Blot, Flow Cytometry
Unconjugated
Purified
TNFRSF11A, RANK
Extracellular domain of human RANK (CD265).
66.03 kDa calculated.
25 μg
Stable for 1 year at 2 to 8°C from date of receipt.
Monoclonal