Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD265/RANK Mouse anti-Human, Unconjugated, Clone: 8G1-24, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $380.00

Specifications

Antigen CD265/RANK
Applications Western Blot, Flow Cytometry
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS20741MI
View Documents
MilliporeSigma
MABS2074100UG
100 μg
Each for $380.00
Add to cart
 
MABS20742MI
View Documents
MilliporeSigma
MABS207425UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

CD265/RANK Monoclonal antibody specifically detects CD265/RANK in Human samples. It is validated for Western Blotting, Flow Cytometry.
Specifications

Specifications

CD265/RANK
Unconjugated
Mouse
Tumor necrosis factor receptor superfamily member 11A;Osteoclast differentiation factor receptor;ODFR;Receptor activator of NF-KB;CD265
Extracellular domain of human RANK (CD265).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot, Flow Cytometry
Purified
Human
TNFRSF11A, RANK
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit