Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD266 (TWEAK Receptor) Monoclonal Antibody (ITEM-1), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: LIFE TECHNOLOGIES 12901941

Description
Mouse Monoclonal Antibody
CD266 (TWEAK Receptor) Monoclonal antibody specifically detects CD266 (TWEAK Receptor) in Human samples. It is validated for Flow CytometrySpecifications
CD266 (TWEAK Receptor) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9NP84 | |
TNFRSF12A | |
Affinity chromatography | |
RUO | |
51330 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
ITEM-1 | |
PE | |
TNFRSF12A | |
AI255180; BOS_22618; C87282; CD266; FGF-inducible 14; Fgfrp2; Fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; Fn14; HPIP; Tnfrsf12a; Tumor necrosis factor receptor superfamily member 12A; TWEAKR; TWEAK-R; Tweak-receptor | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title