Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD266 (TWEAK Receptor) Mouse anti-Human, PE, Clone: ITEM-1, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$136.00 - $294.00

Specifications

Antigen CD266 (TWEAK Receptor)
Concentration 5 μL/Test
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-901-941 Non-distribution product as customer accommodation.
View Documents
N/A 25 Tests
Each for $136.00
Add to cart
 
12-901-942 Non-distribution product as customer accommodation.
View Documents
N/A 100 Tests
Each for $294.00
Add to cart
 
Description

Description

Mouse Monoclonal Antibody

CD266 (TWEAK Receptor) Monoclonal antibody specifically detects CD266 (TWEAK Receptor) in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CD266 (TWEAK Receptor)
Flow Cytometry
PE
Mouse
Human
IgG1, kappa
Affinity chromatography
Antibody
5 μL/Test
Monoclonal
Liquid
RUO
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Monoclonal
4° C, store in dark, DO NOT FREEZE!
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit