Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, BUV737, Clone: L128, BD Horizon™

Mouse Monoclonal Antibody

$240.00 - $456.00

Specifications

Antigen CD27
Clone L128
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Ultraviolet 737
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB612830
View Documents
BD Biosciences
612830
25 Tests
Each for $240.00
Add to Cart
 
BDB612829
View Documents
BD Biosciences
612829
100 Tests
Each for $456.00
Add to Cart
 
Description

Description

CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD27
Flow Cytometry
Brilliant Ultraviolet 737
RUO
IgG1
Refrigerate (2 to 8°C)
L128
Monoclonal
Mouse
Human
Primary
CD27
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit