Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, BV711, Clone: M-T271, BD Horizon™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564893

Catalog No. BDB564893


Add to cart

Description

Description

CD27 Monoclonal antibody specifically detects CD27 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
Specifications

Specifications

CD27
M-T271
Vol. Per test: 5μL
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV711 under optimum conditions
Monoclonal
Flow Cytometry
Brilliant Violet 711
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152
Human T-CLL cells
100 Tests
Cell Biology
Primary
Baboon, Cynomolgus, Human, Rhesus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.