Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Mouse anti-Human, Clone: LPFS2/1611, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14899

Catalog No. 89-034-982


Add to cart

Description

Description

Mouse monoclonal antibody raised against recombinant human CD27.

  • High specificity
  • High reactivity
Specifications

Specifications

CD27
LPFS2/1611
Mouse monoclonal antibody raised against recombinant human CD27.
In 10 mM PBS (0.05% BSA and 0.05% azide).
MGC20393|S152|T14|TNFRSF7|Tp55
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Recognizes a protein of a disulfide-linked 120kDa dimer which identified as CD27.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CD27
Recombinant protein corresponding to human CD27 (exact sequence is proprietary).
Protein A/G purification
RUO
Primary
939
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)