Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ CD27, Rat, Unlabeled, Clone: RM27-3E5,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ 041475I
Description
CD27 antigen (UniProt P41272; also known as CD27, CD27L receptor, T-cell activation antigen CD27, Tumor necrosis factor receptor superfamily member 7) is encoded by the Cd27 (also known as Tnfrsf7) gene (Gene ID 21940) in murine species. CD27 is a TNF receptor superfamily member that binds to CD70 and mediates the survival, function, and differentiation of T-, B-, natural killer (NK), and plasma cells. Interaction with CD70 provides costimulation in T cells induces signaling events essential for cell proliferation, long-term maintenance of antigen-specific T cells, antiviral responses, antitumor immunity, and alloreactivity. CD27 ligation on B-cells by CD70 causes enhanced plasma cell formation and increased IgG production. CD27 is also used as a marker for memory B cells in the classification of B-cell deficiencies, including common variable immunodeficiency (CVID). CD27 is synthesized with a signal peptide (a.a. 1-23), which was removed posttranslationally to yield the mature type I transmembrane protein with an extracellular (a.a. 24-182) composed of three TNFR-Cys repeats (a.a. 26-63, 64-104, 105-141), followed by a transmembrane region (a.a. 183-203), and a cytoplasmic tail (a.a. 204-250).Specifications
CD27 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified rat IgG2a in PBS without preservatives. | |
P41272 | |
Human IgG1 Fc-tagged recombinant mouse CD27 extracellular domain (Sumi, T., et al. (2008). Immunol. Lett. 119(1-2):91-96). | |
100 μL | |
Inflammation & Immunology | |
NP_001028298 | |
Mouse | |
Purified |
Neutralization | |
RM27-3E5 | |
Unconjugated | |
Cd27, Tnfrsf7 | |
Rat | |
Protein G Purified | |
RUO | |
Primary | |
Clone RM27-3E5 enhanced anti-CD3-stimulated T-cell proliferation when immobilized in vitro and augmented OVA-specific CTL induction when administrated in vivo (Roberts, D.J., et al. (2010). J. Immunother. 33(8): 769 779; Sumi, T., et al. (2008). Immunol. Lett. 119(1-2):91-96). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.