Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7, Armenian Hamster anti-Mouse, Clone: LG.3A10, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
$265.00 - $566.50
Specifications
Antigen | CD27/TNFRSF7 |
---|---|
Clone | LG.3A10 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional |
Applications | Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay |
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional.Specifications
CD27/TNFRSF7 | |
1 mg/mL | |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Functional Assay | |
Unconjugated | |
Armenian Hamster | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity | |
PBS | |
939 | |
This antibody was produced by injecting Armenian hamsters intraperitoneally with a mixture of two murine CD27/TNFRSF7 tag-expressing Armenian hamster fibroblast clones (AR0524 and AR0530) in Freund's adjuvant | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
LG.3A10 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Functional | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
CD27 | |
IgG κ | |
Protein A purified | |
This antibody is specific for mouse CD27/TNFRSF7, which, as in the human system, is largely restricted to cells of the T cell lineage, and is expressed on the great majority of both alpha-beta and gamma-delta T lymphocytes, and thymocytes. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title