Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Mouse anti-Human, PE-Cyanine7, Clone: LPFS2/1611, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254582PECY7

Catalog No. N254582PEC7


Add to cart

Description

Description

CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD27/TNFRSF7
LPFS2/1611
Flow Cytometry
Conjugated, Purified
CD27
Recombinant human CD22 protein (exact sequence is proprietary)
Protein A or G purified
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity
Primary
939.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit