Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Rabbit anti-Mouse, Alexa Fluor 594, Clone: 12, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290423AF594

Catalog No. NB039254


Add to cart

Description

Description

CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications

Specifications

CD27/TNFRSF7
12
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity
Primary
939
Flow Cytometry, Immunofluorescence
Alexa Fluor 594
50mM Sodium Borate
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Recombinant Mouse CD27/TNFRSF7 protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit