Learn More
CD272 (BTLA) Mouse anti-Human, BV480, Clone: J168-540, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 746759
Catalog No. BDB746759
Add to cart
Description
Description
CD272 (BTLA) Monoclonal antibody specifically detects CD272 (BTLA) in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD272 (BTLA)
J168-540
Brilliant Violet 480
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human