Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274 (PD-L1, B7-H1) Monoclonal Antibody (MIH5), NovaFluor™ Yellow 700, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH5
Concentration 0.1 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
M036T02Y06
Encompass
View Documents
LIFE TECHNOLOGIES
M036T02Y06
25 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M036T03Y06
Encompass
View Documents
LIFE TECHNOLOGIES
M036T03Y06
100 μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

CD274 (PD-L1, B7-H1) Monoclonal antibody specifically detects CD274 (PD-L1, B7-H1) in Mouse samples. It is validated for Flow Cytometry
Specifications

Specifications

CD274 (PD-L1, B7-H1)
0.1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1
IgG2a λ
Affinity chromatography
Antibody
MIH5
Flow Cytometry
NovaFluor Yellow 700
Rat
Mouse
Q9EP73
60533
Primary
4° C, store in dark, DO NOT FREEZE!
Cd274
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit