Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD274 (PD-L1) Rat anti-Mouse, BV480, Clone: 10F.9G2, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 568589
This item is not returnable.
View return policy
Description
CD274 (PD-L1) Monoclonal antibody specifically detects CD274 (PD-L1) in Mouse samples. It is validated for Flow cytometry (Routinely Tested)Specifications
CD274 (PD-L1) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
60533 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Flow Cytometry | |
10F.9G2 | |
Brilliant Violet 480 | |
Cd274; Pdl1; Pdcd1l1; Pdcd1lg1; B7 homolog 1; B7h1 | |
Mouse PD-L1 | |
0.1 mg | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title