Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD277/BTN3A1 Mouse anti-Human, Clone: BT3.1, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | CD277/BTN3A1 |
---|---|
Clone | BT3.1 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 |
Applications | Flow Cytometry, Immunoprecipitation |
Description
CD277/BTN3A1 Monoclonal specifically detects CD277/BTN3A1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD277/BTN3A1 | |
0.5 mg/mL | |
Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
RUO | |
PBS, pH 7.2 with 0.09% Sodium Azide | |
11119 | |
The immunogen for this antibody is CD277 (B7 family, Butyrophilin). | |
Primary | |
Store at 4C. Do not freeze. |
BT3.1 | |
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Mouse | |
Human | |
butyrophilin, subfamily 3, member A1, dJ45P21.3 (butyrophilin, subfamily 3, member A1) | |
BTN3A1 | |
IgG1 | |
Affinity Purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title