Learn More
CD277 Mouse, anti-Human, R718, Clone: 232-5, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752207
Catalog No. BDB752207
Add to cart
Description
Description
The 232-5 monoclonal antibody specifically recognizes Butyrophilin 3 (BTN3), which is also known as CD277. BTN3 is comprised of three structurally related members, BTN3A1, BTN3A2, and BTN3A3. BTN3 molecules are transmembrane type 1 glycoproteins that belong to the Ig superfamily. The 232-5 antibody recognizes the variable region of the extracellular domains of BTN3 molecules. BTN3 molecules are differentially expressed by T, NKT, B, and NK cells, monocytes, macrophages, dendritic cells, as well as some stem cells and tumor cells. BTN3 expression is decreased upon lymphocyte activation. The extracellular domain of CD277 is structurally related to the B7 family of proteins that regulate T cell activation. A soluble form of the 232-5 antibody can serve as an agonist in BTN3-mediated regulation of anti-CD3 ± anti-CD28 antibody activated CD4+ or CD8+ T cells.
Specifications
Specifications
CD277
232-5
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human