Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD278 (ICOS) Armenian Hamster anti-Mouse, Rat, Rhesus Monkey, PE-Cyanine5, Clone: C398.4A, eBioscience™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen 15994981

 View more versions of this product

Catalog No. 50-141-58


Add to cart

Description

Description

Description: The C398.4A monoclonal antibody reacts with ICOS (Inducible Costimulatory molecule), also known as H4/CRP-1/AILIM, and cross-reacts with mouse and rat ICOS. ICOS is a T cell specific activation molecule and a third member of the CD28/CTLA-4 family. A dimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1, is thought to play a protective role in inflammatory autoimmune diseases and be involved in the development of Th2 cells. C398.4A is reported to display strong costimulation of proliferation of T cell clones, intermediate costimulation on activated T cells, and weak costimulation on fresh resting T cells. It is a good costimulator of IL-10 production especially from pre-activated cells that express high level of H4/ICOS. The epitope recognized by C398.4A is different from that bound by the ICOSL (i.e., C398.4A does not block binding of ICOSL). Applications Reported: The C398.4A antibody has been reported for use in flow cytometric analysis. Applications Tested: This C398.4A antibody has been tested by flow cytometric analysis of Con A-stimulated mouse splenocytes. This can be used at less than or equal to 0.06 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL.

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 667 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered. CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
Specifications

Specifications

CD278 (ICOS)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9WVS0, Q9R1T7
AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat, Rhesus Monkey
Flow Cytometry
C398.4A
PE-Cyanine5
Icos
Liquid
Icos
IgG
50 μg
4° C, store in dark, DO NOT FREEZE!
Primary
54167, 64545, 705788
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.