Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD278 Rat anti-Mouse, BB515, Clone: 7E.17G9, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566214
Description
CD278 Monoclonal antibody specifically detects CD278 in Mouse samples. It is validated for Flow cytometry.Specifications
CD278 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
7E.17G9 | |
BB515 | |
ICOS; Ailim; CCLP; CRP-1; Ly115; Inducible T-cell costimulator | |
Mouse Icos cDNA and ICOS hexahistidine fusion protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
Rat IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.