Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human, Alexa Fluor 488, Clone: EH12.1 (also known as EH12), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 567397
This item is not returnable.
View return policy
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow CytometrySpecifications
CD279 (PD-1) | |
Monoclonal | |
Alexa Fluor 488 | |
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2; hSLE1 | |
Human PD-1 Recombinant Protein | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
EH12.1 (also known as EH12) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title