Learn More
CD279 (PD-1) Mouse anti-Human, Brilliant Ultraviolet 737, Clone: EH12.1 (also known as EH12), BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612791
 View more versions of this product
Catalog No. BDB612791
Add to cart
Description
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD279 (PD-1)
EH12.1 (also known as EH12)
Aqueous buffered solution containing ≤0.09% sodium azide.
PD1; hPD-1; hPD-l; PDCD1; PDC1; Programmed cell death 1; SLEB2
Human PD-1 Recombinant Protein
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 737
Affinity Purified
Mouse
IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal