Learn More
Novus Biologicals™ CD28 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191279
Catalog No. NBP191279
Add to cart
Description
Description
The Human CD28 ELISA Kit (Colorimetric) measures human CD28 in cell culture supernatant, serum.
Specifications
Specifications
0.78 - 50ng/mL
940
ELISA Kit
0.18ng/mL
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Apoptosis, CD Markers, Immunology
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD28, Biotin-Conjugate anti-human sCD28 monoclonal antibody, Streptavidin-HRP, Human sCD28 Standard lyophilized, 100ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
CD28
CD28