Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Golden Syrian Hamster anti-Mouse, Phycoerythrin, Clone: 37.51, Novus Biologicals™

Golden Syrian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP130418PE

Catalog No. NBP130418PE


Add to cart

Description

Description

CD28 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen).
Specifications

Specifications

CD28
37.51
Flow Cytometry
Purified
CD28
Mouse T cell lymphoma EL-4 cells
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
The Syrian hamster monoclonal antibody 37.51 reacts with extracellular domain of mouse CD28 costimulatory receptor, a 45 kDa homodimeric protein expressed by thymocytes, mature T cells and NK cells.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Golden Syrian Hamster
IgG
0.1 ml
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit