Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Syrian Hamster anti-Mouse, Clone: 37.51.1, Invitrogen™ Non-distribution product as customer accommodation.

Syrian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen HM35005

Catalog No. 50-113-8079


Add to cart

Description

Description

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. CD28 is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes. Moreover, CD28 is the critical T cell costimulatory receptor that provides the cell the important second activation signal by binding CD80 and CD86 which are expressed by antigen presenting cells. In addition to its co-stimulation role, CD28 functions by preventing T cells from entering an anergic-hyporesponsive state or from undergoing premature apoptotic cell death. In murine peripheral lymphoid organs and in the blood, all CD4+ and CD8+ T cells express CD28. In the thymus, CD28 expression is highest on immature CD3-, CD8+ and CD4+8+ cells, and on CD4-8- cells that express alpha/beta and gamma/delta TCR. The level of CD28 on mature CD4+ and CD8+ alpha/beta TCR+ thymocytes is two- to fourfold lower than on the immature cells. Diseases associated with CD28 dysfunction include mycosis fungiodes and Sezary’s Disease.
Specifications

Specifications

CD28
Monoclonal
Unconjugated
Cd28
Liquid
Cd28
Mouse T lymphoma EL-4 cells.
Purified
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
37.51.1
PBS with 0.1% sodium azide
P31041
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Syrian Hamster
IgG
2.5 mL
4° C
Primary
12487
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit