Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28 Monoclonal Antibody (OTI8C4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF813689


Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow CytometrySpecifications
CD28 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P10747 | |
Mouse | |
Protein A/G | |
RUO | |
940 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry | |
OTI8C4 | |
Unconjugated | |
Cd28 | |
antigen CD28; CD28; CD28 antigen; CD28 antigen (Tp44); CD28 isoform; CD28 isoform 2; Cd28 molecule; CD28 precursor protein; CD28 protein; CD28 protein precursor; CD28RNA; cell surface protein; CHT28; contains partial extracellular domain; costimulatory molecule B7 receptor CD28; EGK_04709; MGC138290; sCD28; soluble CD28; T44; T-cell costimulatory molecule CD28; T-cell costimulatory molecule Tp44; T-cell-specific surface glycoprotein CD28; T-cell-specific surface glycoprotein CD28 homolog; TP44 | |
Full length human recombinant protein of human CD28 (NP_006130) produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction