Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, Clone: C28/1636, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14703

Catalog No. 89-034-786


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human CD28.

  • High specificity
  • High reactivity
Specifications

Specifications

CD28
C28/1636
Mouse monoclonal antibody raised against full length recombinant human CD28.
In 10 mM PBS (0.05% BSA and 0.05% azide).
MGC138290|Tp44
Mouse
IgG1
100 ug
Store at 4°C.
Monoclonal
Recognizes a glycoprotein of 44-88kDa which is identified as CD28.
Flow Cytometry, Immunofluorescence
Unconjugated
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CD28
Recombinant protein corresponding to full-length human CD28.
Protein A/G purification
RUO
Primary
940
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)