Learn More
CD28 Mouse anti-Human, Janelia Fluor 549, Clone: 204.12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142206JF549
Catalog No. NBP42206549
Add to cart
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD28
204.12
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD28
The antibody reacts with the CD28 antigen.
Protein A or G purified
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Human, Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Mouse
IgG2a
0.1mL
Store at 4°C in the dark.
Monoclonal
204-12 remained unassigned at the VI international workshop on human leucocyte differentiation antigens. Source: A BALB/c mouse was immunized with stimulated human leucocytes. Splenocytes were fused with NS1.