Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse anti-Human, PE/Cy5.5, Clone: C28/1636, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254573PECY55

Catalog No. N254573PEC5


Add to cart

Description

Description

CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD28
C28/1636
Flow Cytometry
Conjugated, Purified
CD28
Recombinant human full-length CD28 protein
Protein A or G purified
Adaptive Immunity, Apoptosis, Immunology
Primary
940.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit