Learn More
CD28 Monoclonal Antibody (CD28.2), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$341.00
Specifications
Antigen | CD28 |
---|---|
Clone | CD28.2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3276
|
Invitrogen
25028942 |
100 Tests |
Each of 1 for $341.00
|
|
|||||
Description
Description: The CD28.2 monoclonal antibody reacts with the human CD28 molecule, a 44 kDa homodimer expressed by thymocytes, mature T cells and plasma cells. CD28 is a ligand for CD80 (B7-1) and CD86 (B7-2) and is a potent co-stimulator of T cells. Signaling through CD28 augments IL-2 and IL-2 receptor expression as well as cytotoxicity of CD3-activated T cells. Applications Reported: This CD28.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CD28.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.
CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Flow Cytometry.Specifications
CD28 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P10747 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
CD28.2 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
940 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.