Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Mouse, Biotin, Clone: 204.12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142206B

Catalog No. NBP142206B


Add to cart

Description

Description

CD28 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications

Specifications

CD28
204.12
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD28
The antibody reacts with the CD28 antigen.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
204-12 remained unassigned at the VI international workshop on human leucocyte differentiation antigens. Source: A BALB/c mouse was immunized with stimulated human leucocytes. Splenocytes were fused with NS1.
Immunohistochemistry, Immunohistochemistry (Frozen)
Biotin
PBS with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Mouse
IgG2a
0.1 ml
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit