Learn More
CD28 Mouse, HRP, Clone: 204.12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142206H
Catalog No. NBP142206H
Add to cart
Description
Description
CD28 Monoclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
CD28
204.12
Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD28
The antibody reacts with the CD28 antigen.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
204-12 remained unassigned at the VI international workshop on human leucocyte differentiation antigens. Source: A BALB/c mouse was immunized with stimulated human leucocytes. Splenocytes were fused with NS1.
Immunohistochemistry, Immunohistochemistry (Frozen)
HRP
PBS with No Preservative
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Mouse
IgG2a
0.1 ml
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Human, Mouse